EMAIL THIS PAGE TO A FRIEND
Photodermatology, photoimmunology & photomedicine
UVA irradiation of the eye modulates the contact hypersensitivity of the skin and intestines by affecting mast cells in mice.
PMID 25495784
Abstract
Ultraviolet A (UVA) irradiation before allergic sensitization induces immunosuppression, but the precise mechanism remained unclear. In this study, we examined the influence of UVA irradiation of the eye on contact hypersensitivity (CHS) and the role of mast cells in CHS. We used two types of haptens, fluorescein isothiocyanate (FITC: a Th2 type hapten) and 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one (oxazolone: a Th1 type hapten). A 300 kJ/m(2) dose of UVA irradiation was delivered to the eyes. After UVA irradiation, we sensitized abdominal shaved skin and challenged the ear epidermis and colons of these mice with each hapten. After UVA irradiation, the CHS of the skin and colon were not inhibited in the FITC-sensitized mice. However, in the oxazolone-sensitized mice, only the CHS of the skin was inhibited by UVA irradiation. The inflammation of the colon became more severe after UVA irradiation. In mast cell-deficient (W/Wv) mice sensitized to FITC, the CHS was weaker than that in WT mice. Moreover, the reduction of immunosuppression in ear swelling was seen for one of the two models they used. These results suggest that the mast cells induced by UVA irradiation of the eye have different roles in the epidermis and colon and have different responses to different haptens.