EMAIL THIS PAGE TO A FRIEND
Malaria journal
Microsatellite markers reveal low levels of population sub-structuring of Plasmodium falciparum in southwestern Nigeria.
PMID 25496185
Abstract
Genetic diversity studies provide evidence of Plasmodium falciparum differentiation that could affect fitness and adaptation to drugs and target antigens for vaccine development. This study describes the genetic structure of P. falciparum populations in urban and rural sites from southwestern Nigeria. Ten neutral microsatellite loci were genotyped in 196 P. falciparum infections from three localities: Aramoko-Ekiti, a rural community; Lekki, an urban location and Badagry, a peri-urban border settlement. Analysis was performed on the genetic diversity, linkage disequilibrium, population structure and inter-population differentiation. Allelic diversity values were similar across all populations, with mean expected heterozygosity (HE) values between 0.65 and 0.79. No matching multilocus haplotypes were found and analysis of multilocus LD showed no significant index of association. Genetic differentiation between populations was low (ΦPT = 0.017). The absence of detectable population structure of P. falciparum in southwestern Nigeria is evident in the lack of significant differentiation between populations separated by about 200 km. This implies that a fairly uniform malaria control strategy may be effective over a wide geographic range in this highly endemic region. However, more wide-scale survey across the country will be required to inform malaria control in this large and densely populated endemic region.