EMAIL THIS PAGE TO A FRIEND
Histopathology
Frequent co-inactivation of the SWI/SNF subunits SMARCB1, SMARCA2 and PBRM1 in malignant rhabdoid tumours.
PMID 25496315
Abstract
Malignant rhabdoid tumours (MRTs) are highly aggressive malignancies of early infancy characterized by inactivation of SMARCB1, a core member of the SWI/SNF chromatin-remodelling complex. The aim of this study was to explore the status of multiple key subunits of the SWI/SNF complex in MRTs. We screened the key subunits of the SWI/SNF complex, including SMARCB1, SMARCA2, PBRM1, SMARCA4, and ARID1A, in four MRTs by immunohistochemistry, sequencing, and fluorescence in-situ hybridization (FISH). Complete loss of SMARCB1, SMARCA2 and PBRM1 expression and corresponding mutations in the same genes were observed in all cases. The mutations included seven missense, three same-sense, four frameshift and two truncating mutations. FISH revealed heterozygous deletion of SMARCB1 in one case, and monoploidy of chromosome 22, which harbours SMARCB1, in another case. Furthermore, trisomy of chromosome 9, which harbours SMARCA2, was observed in two cases. Abnormality of PBRM1 was not found in any case. We report, for the first time, co-inactivation and frequent mutations of SMARCB1, SMARCA2 and PBRM1 in MRTs. Multiple subunit abnormalities of the SWI/SNF complex potentially act together to contribute to the tumorigenesis of MRTs, which provides unique insights into this disease.