EMAIL THIS PAGE TO A FRIEND
Natural product research
Fast determination of Ziziphora tenuior L. essential oil by inorganic-organic hybrid material based on ZnO nanoparticles anchored to a composite made from polythiophene and hexagonally ordered silica.
PMID 25496469
Abstract
In this paper, for the first time, an inorganic-organic hybrid material based on ZnO nanoparticles was anchored to a composite made from polythiophene and hexagonally ordered silica (ZnO/PT/SBA-15) for use in solid-phase fibre microextraction (SPME) of medicinal plants. A homemade SPME apparatus was used for the extraction of volatile components of Ziziphora tenuior L. A simplex method was used for optimisation of five different parameters affecting the efficiency of the extraction. The main constituents extracted by ZnO/PT/SBA-15 and PDMS fibres and hydrodistillation (HD) methods, respectively, included pulegone (51.25%, 53.64% and 56.68%), limonene (6.73%, 6.58% and 8.3%), caryophyllene oxide (5.33%, 4.31% and 4.53%) and 1,8-cineole (4.21%, 3.31% and 3.18%). In comparison with the HD method, the proposed technique could equally monitor almost all the components of the sample, in an easier way, in a shorter time and requiring a much lower amount of the sample.