EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Biodistribution of nanostructured lipid carriers: a tomographic study.
PMID 25497177
Abstract
This study describes the preparation, characterization, and biodistribution of radiolabelled nanostructured lipid carriers (NLCs) especially designed for in vivo tomographic study. A preliminary formulative study was conducted in order to incorporate (99m)Tc based tracer in NLCs. At this aim a (99m)Tc complex containing a terminal (99m)Tc ≡ N multiple bond ([(99m)Tc]N-DBODC2) has been synthesized and included in NLCs produced by a stirring and ultrasonication method. The morphological and dimensional characteristics of the produced NLCs have been accurately investigated by a number of specific techniques, including: cryogenic transmission electron microscopy, X-ray, photon correlation spectroscopy and sedimentation field flow fractionation. The obtained NLCs were employed for achieving in vivo tomographic images of the rat body by small-animal SPECT scanner that enabled the investigation of NLC biodistribution after intraperitoneal, intravenous, intranasal and oral administration. NLC production protocol allowed to firmly encapsulate the radiotracer within the nanoparticles. In vivo studies evidenced that NLC remained stable in vivo, suggesting their suitability as controlled release system for drugs and radiochemical for therapeutic and diagnostic purposes. Moreover the high resolution images obtained by the SPECT technique allowed to detect NLC presence in brown fat tissue, suggesting NLC therapeutic application for treating human obesity and related metabolic disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5502
Succinic dihydrazide, 96%
C4H10N4O2