EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Spontaneously hyperactive MEK-Erk pathway mediates paradoxical facilitation of cell proliferation in mild hypoxia.
PMID 25497211
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6416
GW5074, powder
C15H8Br2INO2