EMAIL THIS PAGE TO A FRIEND
International Immunopharmacology
Optimisation of protein microarray techniques for analysis of the plasma proteome: minimisation of non-specific binding interactions.
PMID 25497232