EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Analyte quantification with comprehensive two-dimensional gas chromatography: assessment of methods for baseline correction, peak delineation, and matrix effect elimination for real samples.
PMID 25497580