EMAIL THIS PAGE TO A FRIEND
Colloids and Surfaces. B, Biointerfaces
A new strategy for synthesizing AgInS? quantum dots emitting brightly in near-infrared window for in vivo imaging.
PMID 25499228
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
685666
N,N′-Diisopropylcarbodiimide, ChemDose tablets, Loading: 0.15mmol per tablet
C7H14N2