EMAIL THIS PAGE TO A FRIEND
Blood
Mantle cell lymphoma: evolving management strategies.
PMID 25499451
Abstract
Mantle cell lymphoma (MCL) is a rare and aggressive form of non-Hodgkin's lymphoma that generally affects older individuals and continues to have one of the worst outcomes of all the lymphomas. Over the last decade, there has been a widespread adoption of cytarabine-based therapy in younger patients, and the incorporation of rituximab into chemotherapeutic regimens has become an evidence-based standard of care. However MCL remains a largely incurable disease, and following relapse, it can be a challenge to manage. Although it is possible to define prognosis reliably, there are, as yet, no clear diagnostic or response-adjusted parameters that can help to guide therapeutic decisions. However, there are a number of highly active targeted therapies that are moving into the clinic that are set to transform the therapeutic paradigm for this disease in the very near future. This review will explore the molecular pathogenesis of MCL and the current and evolving therapeutic strategies for this disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6645
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
C9H13N3O5 · HCl