EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Analyses of chicken sialyltransferases related to N-glycosylation.
PMID 25499752
Abstract
Proteins exogenously expressed and deposited in the egg whites of transgenic chickens did not contain terminal sialic acid in their N-glycan. Since this sugar is important for the biological stability of therapeutic proteins, we examined chicken sialyltransferases (STs). Based on homologies in DNA sequences, we cloned and expressed several chicken STs, which appeared to be involved in N-glycosylation in mammals, in 293FT cells. Enzymatic activity was detected with ST3Gal3, ST3Gal6 and ST6Gal1 using galactose-β1,4-N-acetylglucosamine (Galβ1,4GlcNAc) as an acceptor. Using Golgi fractions from the cell-free extracts of chicken organs, α2,3- and/or α2,6-ST activities were detected in the liver and kidney, but were absent in the oviduct cells in which egg-white proteins were produced. This result suggested that the lack of ST activities in oviduct cells mainly caused the lack of sialic acid in the N-glycan of proteins exogenously expressed and deposited in egg white.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1131
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
C9H14N3O8P