EMAIL THIS PAGE TO A FRIEND
Retrovirology
Cytotoxic T lymphocyte lysis of HTLV-1 infected cells is limited by weak HBZ protein expression, but non-specifically enhanced on induction of Tax expression.
PMID 25499803