EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
DODAB:monoolein liposomes containing Candida albicans cell wall surface proteins: a novel adjuvant and delivery system.
PMID 25499956