EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Extragenital lichen sclerosus: clinical, dermoscopic, confocal microscopy and histologic correlations.
PMID 25500042
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9879 Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C7806 Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
C1188 Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
C8919 Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
C9791 Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
C3511 Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
C9301 Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
C0543 Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder (from 0.2 μm-filtered solution), BioReagent, suitable for cell culture
C7774 Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
C4407 Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
C8374
Collagen from human placenta, Bornstein and Traub Type IV, powder
H4417 Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
C3657 Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
C5608 Collagen from rabbit skin, Bornstein and Traub Type I, powder
C5483
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
C6745 Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
K0253
Keratin from human epidermis, aqueous solution