EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
DNA-protein immunization using Leishmania peroxidoxin-1 induces a strong CD4+ T cell response and partially protects mice from cutaneous leishmaniasis: role of fusion murine granulocyte-macrophage colony-stimulating factor DNA adjuvant.
PMID 25500571