EMAIL THIS PAGE TO A FRIEND
Chest
Endotracheal tubes for critically ill patients: an in vivo analysis of associated tracheal injury, mucociliary clearance, and sealing efficacy.
PMID 25500677
Abstract
Improvements in the design of the endotracheal tube (ETT) have been achieved in recent years. We evaluated tracheal injury associated with ETTs with novel high-volume low-pressure (HVLP) cuffs and subglottic secretions aspiration (SSA) and the effects on mucociliary clearance (MCC). Twenty-nine pigs were intubated with ETTs comprising cylindrical or tapered cuffs and made of polyvinylchloride (PVC) or polyurethane. In specific ETTs, SSA was performed every 2 h. Following 76 h of mechanical ventilation, pigs were weaned and extubated. Images of the tracheal wall were recorded before intubation, at extubation, and 24 and 96 h thereafter through a fluorescence bronchoscope. We calculated the red-to-green intensity ratio (R/G), an index of tracheal injury, and the green-plus-blue (G+B) intensity, an index of normalcy, of the most injured tracheal regions. MCC was assessed through fluoroscopic tracking of radiopaque markers. After 96 h from extubation, pigs were killed, and a pathologist scored injury. Cylindrical cuffs presented a smaller increase in R/G vs tapered cuffs (P = .011). Additionally, cuffs made of polyurethane produced a minor increase in R/G (P = .012) and less G+B intensity decline (P = .022) vs PVC cuffs. Particularly, a cuff made of polyurethane and with a smaller outer diameter outperformed all cuffs. SSA-related histologic injury ranged from cilia loss to subepithelial inflammation. MCC was 0.9 ± 1.8 and 0.4 ± 0.9 mm/min for polyurethane and PVC cuffs, respectively (P < .001). HVLP cuffs and SSA produce tracheal injury, and the recovery is incomplete up to 96 h following extubation. Small, cylindrical-shaped cuffs made of polyurethane cause less injury. MCC decline is reduced with polyurethane cuffs.