EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Uncoupling AMPK from autophagy: a foe that hinders the beneficial effects of metformin treatment on metabolic syndrome-associated atherosclerosis? Focus on "glucose and palmitate uncouple AMPK from autophagy in human aortic endothelial cells".
PMID 25500743