EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Dual inhibition of Bcr-Abl and Hsp90 by C086 potently inhibits the proliferation of imatinib-resistant CML cells.
PMID 25501124
Abstract
Although tyrosine kinase inhibitors (TKI) such as imatinib provide an effective treatment against Bcr-Abl kinase activity in the mature cells of patients with chronic myelogenous leukemia (CML), TKIs probably cannot eradicate the leukemia stem cell (LSC) population. Therefore, alternative therapies are required to target both mature CML cells with wild-type (WT) or mutant Bcr-Abl and LSCs. To investigate the effect of C086, a derivative of curcumin, on imatinib-resistant cells, we explored its underlying mechanisms of Bcr-Abl kinase and heat shock protein 90 (Hsp90) function inhibition. Biochemical assays were used to test ABL kinase activity; fluorescence measurements using recombinant NHsp90, Hsp90 ATPase assay, immunoprecipitation, and immunoblotting were applied to examine Hsp90 function. Colony-forming unit, long-term culture-initiating cells (LTC-IC), and flow cytometry were used to test CML progenitor and stem cells. Biochemical assays with purified recombinant Abl kinase confirmed that C086 can directly inhibit the kinase activity of Abl, including WT and the Q252H, Y253F, and T315I mutations. Furthermore, we identified C086 as a novel Hsp90 inhibitor with the capacity to disrupt the Hsp90 chaperone function in CML cells. Consequently, it inhibited the growth of both imatinib-sensitive and -resistant CML cells. Interestingly, C086 has the capacity to inhibit LTC-ICs and to induce apoptosis in both CD34(+)CD38(+) and CD34(+)CD38(-) cells in vitro. Moreover, C086 could decrease the number of CD45(+), CD45(+)CD34(+)CD38(+), and CD45(+)CD34(+)CD38(-) cells in CML NOD-SCID mice. Dual suppression of Abl kinase activity and Hsp90 chaperone function by C086 provides a new therapeutic strategy for treating Bcr-Abl-induced leukemia resistant to TKIs.