EMAIL THIS PAGE TO A FRIEND
PLoS Genetics
Mutations in global regulators lead to metabolic selection during adaptation to complex environments.
PMID 25501822
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7408
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1