EMAIL THIS PAGE TO A FRIEND
Cell death & disease
A loss of FUS/TLS function leads to impaired cellular proliferation.
PMID 25501833
Abstract
Fused in sarcoma/translocated in liposarcoma (FUS/TLS or FUS) is a multifunctional RNA/DNA-binding protein that is pathologically associated with cancer and neurodegeneration. To gain insight into the vital functions of FUS and how a loss of FUS function impacts cellular homeostasis, FUS expression was reduced in different cellular models through RNA interference. Our results show that a loss of FUS expression severely impairs cellular proliferation and leads to an increase in phosphorylated histone H3, a marker of mitotic arrest. A quantitative proteomics analysis performed on cells undergoing various degrees of FUS knockdown revealed protein expression changes for known RNA targets of FUS, consistent with a loss of FUS function with respect to RNA processing. Proteins that changed in expression as a function of FUS knockdown were associated with multiple processes, some of which influence cell proliferation including cell cycle regulation, cytoskeletal organization, oxidative stress and energy homeostasis. FUS knockdown also correlated with increased expression of the closely related protein EWS (Ewing's sarcoma). We demonstrate that the maladaptive phenotype resulting from FUS knockdown is reversible and can be rescued by re-expression of FUS or partially rescued by the small-molecule rolipram. These results provide insight into the pathways and processes that are regulated by FUS, as well as the cellular consequences for a loss of FUS function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
PHR1125
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H10N2O3
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
81845
Propidium iodide, ≥94% (HPLC)
C27H34I2N4
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N
85886
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
C21H39N7O12 · 1.5H2SO4
S1400000
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
C21H39N7O12 · 1.5H2O4S
S6501
Streptomycin sulfate salt, powder
C21H39N7O12 · 1.5H2O4S
S9137
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
C21H39N7O12 · 1.5H2O4S
S1277
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
C21H39N7O12 · 1.5H2O4S
T3258
Tetracycline, ≥88.0% (HPLC)
C22H24N2O8 · xH2O
87128
Tetracycline, ≥98.0% (NT)
C22H24N2O8 · xH2O
74564
Trifluoroacetic acid, analytical standard
C2HF3O2