EMAIL THIS PAGE TO A FRIEND
International Journal of Oncology
CCL18 promotes epithelial-mesenchymal transition, invasion and migration of pancreatic cancer cells in pancreatic ductal adenocarcinoma.
PMID 25502147