EMAIL THIS PAGE TO A FRIEND
Anticancer Research
Tanshinone IIA inhibits gastric carcinoma AGS cells through increasing p-p38, p-JNK and p53 but reducing p-ERK, CDC2 and cyclin B1 expression.
PMID 25503137