EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Identification of benzenesulfonamide quinoline derivatives as potent HIV-1 replication inhibitors targeting Rev protein.
PMID 25503645
Abstract
Human immunodeficiency virus type 1 (HIV-1) Rev protein facilitates the export of viral RNA from nucleus to cytoplasm, which is a key step in HIV-1 pathogenesis and transmission. In this study, we have screened a commercial library and identified the hit compound 1 bearing a benzenesulfonamide quinoline scaffold that inhibited Rev activity and HIV-1 infectivity. Compounds bearing this scaffold were synthesized and their SAR was studied. We identified compound 20 with low toxicity and potent activity to inhibit HIV-1 replication by affecting Rev function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
548081
2,4-Difluorobenzenesulfonyl chloride, 97%
C6H3ClF2O2S