EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Wee-1 kinase inhibition overcomes cisplatin resistance associated with high-risk TP53 mutations in head and neck cancer through mitotic arrest followed by senescence.
PMID 25504633