EMAIL THIS PAGE TO A FRIEND
Journal of biomedical materials research. Part A
Continuous multilayered composite hydrogel as osteochondral substitute.
PMID 25504681
Abstract
Cartilage is a highly organized avascular soft tissue that assembles from nano-to macro-scale to produce a complex structural network. To mimic cartilage tissue, we developed a stable multilayered composite material, characterized by a tailored gradient of mechanical properties. The optimized procedure implies chemical crosslinking of each layer directly onto the previous one and ensures a drastic reduction of the material discontinuities and brittleness. The multilayered composite was characterized by infrared spectroscopy, differential scanning calorimetry, thermogravimetry, and scanning electron microscopy in order to compare its physico-chemical characteristics with those of cartilage tissue. The rheological behavior of the multilayered composite was similar to that of human cartilage. Finally its cytocompatibility toward chondrocytes and osteoblasts was evaluated.