EMAIL THIS PAGE TO A FRIEND
The Annals of applied biology
Simultaneous detection of major blackleg and soft rot bacterial pathogens in potato by multiplex polymerase chain reaction.
PMID 25506085
Abstract
A multiplex polymerase chain reaction (PCR) assay for simultaneous, fast and reliable detection of the main soft rot and blackleg potato pathogens in Europe has been developed. It utilises three pairs of primers and enables detection of three groups of pectinolytic bacteria frequently found in potato, namely: