EMAIL THIS PAGE TO A FRIEND
The Journal of oral implantology
Retentiveness of various luting agents used with implant-supported prosthesis: an in vitro study.
PMID 25506659
Abstract
Desired retrievability of cemented implant-supported fixed prosthesis makes the retentive strength of cementing agents an important consideration. The aim of the study was to evaluate the retentiveness of purposely designed implant cement and compare its retentiveness with dental cements that are commonly used with implant systems. Ten implant analogs were embedded in auto-polymerizing acrylic resin blocks and titanium abutments were attached to them. Fifty standardized copings were waxed directly on the abutment and casted. The cements used were: (1) resin-bonded zinc oxide eugenol cement, (2) purposely designed implant cement, (3) zinc phosphate cement, (4) zinc polycarboxylate cement, and (5) glass ionomer cement. After cementation, each sample was subjected to a pull-out test using universal testing machine and loads required to remove the crowns were recorded. The mean values and standard deviations of cement failure loads were analyzed using ANOVA and Bonferroni test. The mean values (± SD) of loads at failure (n = 10) for various cements were as follows (N): resin-bonded zinc oxide eugenol cement 394.62 (± 9.76), Premier implant cement 333.86 (± 18.91), zinc phosphate cement 629.30 (± 20.65), zinc polycarboxylate cement 810.08 (± 11.52), and glass ionomer cement 750.17 (± 13.78). The results do not suggest that one cement type is better than another, but they do provide a ranking order of the cements regarding their ability to retain the prosthesis and facilitate easy retrievability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35995
Eugenol, PESTANAL®, analytical standard
C10H12O2
PHR1269
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O2
79891
Eugenol, certified reference material, TraceCERT®
C10H12O2
01050595
Eugenol, primary reference standard
C10H12O2
E5000000
Eugenol, European Pharmacopoeia (EP) Reference Standard
C10H12O2
F1635
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
CH2O
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
348848
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
GF03980932
Titanium, mesh, 150x150mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
267481
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
GF76772391
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Ti
GF24976525
Titanium, rod, 200mm, diameter 25mm, annealed, 99.6+%
Ti
GF90731505
Titanium, tube, 500mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Ti
GF62738006
Titanium, tube, 500mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Ti
GF66180034
Titanium, tube, 500mm, outside diameter 1.05mm, inside diameter 0.75mm, wall thickness 0.15mm, annealed, 99.6+%
Ti
GF86399106
Titanium, tube, 500mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Ti
GF32919925
Titanium, tube, 500mm, outside diameter 10.3mm, inside diameter 8.7mm, wall thickness 0.8mm, annealed, 99.6+%
Ti