EMAIL THIS PAGE TO A FRIEND
Calcified Tissue International
Phosphate binders prevent phosphate-induced cellular senescence of vascular smooth muscle cells and vascular calcification in a modified, adenine-based uremic rat model.
PMID 25511229
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94965
Vinylene carbonate, purum, ≥97.0% (GC)
C3H2O3