EMAIL THIS PAGE TO A FRIEND
International Journal of Biological Macromolecules
Cytotoxicity of curcumin silica nanoparticle complexes conjugated with hyaluronic acid on colon cancer cells.
PMID 25511568
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235768
Vinyltrimethoxysilane, 98%
C5H12O3Si
440221
Vinyltrimethoxysilane, 97%
C5H12O3Si