EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Mechanistic basis of altered morphine disposition in nonalcoholic steatohepatitis.
PMID 25512370
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8777
Morphine sulfate salt pentahydrate
C34H40N2O10S · 5H2O