EMAIL THIS PAGE TO A FRIEND
Journal of the Science of Food and Agriculture
Effects of four different cooking methods on anthocyanins, total phenolics and antioxidant activity of black rice.
PMID 25513670
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52976
Kuromanin chloride, ≥95% (HPLC)
C21H21ClO11
44689
Kuromanin chloride, analytical standard
C21H21ClO11