EMAIL THIS PAGE TO A FRIEND
Nucleic acid therapeutics
Blocking the maturation of OncomiRNAs using pri-miRNA-17∼92 aptamer in retinoblastoma.
PMID 25513843
Abstract
The miR-17∼92. or oncomiR-1, cluster encodes oncogenic microRNAs (miRNAs), and it also promotes retinoblastoma (RB) tumor formation. Antagomir and miRNA mimics based approaches are widely tried against oncogenic and tumor suppressive miRNAs. Other methods for targeting cancer related miRNAs are still under development. In the current study, we focused on the pri-miRNA-17∼92 aptamer (pri-apt), which can potentially replace the mix of five antagomirs by one aptamer that function to abrogate the maturation of miR-17, miR-18a, and miR-19b (P<0.05) for targeting RB. We used RB cell lines WERI-Rb1 and Y79 as an in vitro model. Cellular changes upon transfecting the pri-apt led to S-phase arrest in WERI-Rb1 cells and onset of apoptosis in both Y79 and WERI-Rb1 cell lines. There was increased cytotoxicity as measured by lactate dehydrogenase activity in pri-apt treated Y79 cells (P<0.05), and significant inhibition of cell proliferation was observed in both of the cell lines. Thus we showed the antiproliferative property of pri-apt in RB cell lines, which can be readily modified by developing appropriate vectors for the delivery of the aptamer specifically to cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CYTODET-RO
Cytotoxicity Detection KitPLUS (LDH)