EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
Micelle formulation of hexadecyloxypropyl-cidofovir (HDP-CDV) as an intravitreal long-lasting delivery system.
PMID 25513956