EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Simultaneous targeting of two ligand-binding sites on VEGFR2 using biparatopic Affibody molecules results in dramatically improved affinity.
PMID 25515662