EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Quantification of endogenous metabolites by the postcolumn infused-internal standard method combined with matrix normalization factor in liquid chromatography-electrospray ionization tandem mass spectrometry.
PMID 25516166