EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
In vitro and in vivo antimicrobial efficacy of natural plant-derived compounds against Vibrio cholerae of O1 El Tor Inaba serotype.
PMID 25516242
Abstract
In this study, we investigated antibacterial activities of 20 plant-derived natural compounds against Gram-negative enteric pathogens. We found that both flavonoids and non-flavonoids, including honokiol and magnolol, possess specific antibacterial activities against V. cholerae, but not against other species of Gram-negative bacterium which we tested. Using various antibacterial assays, we determined that there was a dose-dependent bactericidal and biofilm inhibitory activity of honokiol and magnolol against Vibrio cholerae. In addition to antibacterial activities, these molecules also induced an attenuating effect on reactive oxygen species (ROS) production and pro-inflammatory responses generated by macrophages in response to lipopolysaccharides (LPS). Additionally, Caenorhabditis elegans lethality assay revealed that honokiol and magnolol have an ability to extend a lifespan of V. cholerae-infected worms, contributing to prolonged survival of worms after lethal infection. Altogether, our data show for the first time that honokiol and magnolol may be considered as attractive protective or preventive food adjuncts for cholera.