EMAIL THIS PAGE TO A FRIEND
The Journal of Adhesive Dentistry
Microtensile bond strength of lithium disilicate ceramics to resin adhesives.
PMID 25516886
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339261
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
HF
30107
Hydrofluoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥48%
HF
30103
Hydrofluoric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥40%
HF
01066
Hydrofluoric acid, technical, 40-45%
HF
695068
Hydrofluoric acid, ACS reagent, 48%
HF
34877
Water, HPLC Plus
H2O
270733
Water, for HPLC
H2O
320072
Water, ACS reagent
H2O
38796
Water, deionized
H2O
419141
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zr
403296
Zirconium, powder, −100 mesh
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr
267651
Zirconium, sponge, ≥99% trace metals basis
Zr
756385
Zirconium, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf)
Zr
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF27504402
Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF13629483
Zirconium, foil, 50x50mm, thickness 0.1mm, annealed, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04051886
Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%
Zr
GF14815593
Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%
Zr
GF58113019
Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%
Zr
GF67870468
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zr
GF58442070
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF75562869
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF55287221
Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%
Zr
GF38009044
Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF89139594
Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%
Zr