EMAIL THIS PAGE TO A FRIEND
Organic letters
Silver-catalyzed C-H trifluoromethylation of arenes using trifluoroacetic acid as the trifluoromethylating reagent.
PMID 25517200
Abstract
Direct trifluoromethylation of arenes using TFA as the trifluoromethylating reagent was achieved with Ag as the catalyst. This reaction not only provides a new protocol for aryl C-H trifluoromethylation, but the generation of CF3· from TFA may prove useful in other contexts and could potentially be extended to other trifluoromethylation reactions.