EMAIL THIS PAGE TO A FRIEND
Clinical oral investigations
Periodontal treatment outcomes during pregnancy and postpartum.
PMID 25518813
Abstract
This study was conducted to compare periodontal therapy outcomes during pregnancy and after delivery. One hundred nine pregnant women up to the 20th gestational week (GW) were randomized into two groups: the test group (comprehensive periodontal therapy during pregnancy) and the control group (comprehensive periodontal therapy after delivery). Periodontal examinations comprised plaque index (PI), gingival index (GI), periodontal probing depth (PPD), clinical attachment level (CAL), bleeding on probing (BOP), and gingival crevicular fluid (GCF) volume. After baseline examination, women in the test group received periodontal treatment up to the 24th GW. The final examination was performed at the 26th to the 28th GW. Women in the control group were treated 30xa0days after delivery and reexamined 30xa0days after treatment. Periodontal therapy significantly reduced periodontal inflammation in both groups. The mean percentage of sites with BOP was reduced from 49.14xa0% (±22.49) to 11.10xa0% (±7.84) and from 45.71xa0% (±17.86) to 8.07xa0% (±5.21) in the test and control groups, respectively (p = 0.95). No statistically significant differences were observed between groups concerning PI, GI, PPD, CAL, and GCF. The reduction in mean percentage of sites with BOP stratified for initial PPD ≥4xa0mm was higher in the control group (p < 0.01), but no differences were seen regarding GCF in these sites. Hormonal changes during pregnancy do not interfere in treatment outcomes in women with widespread gingival inflammation and limited periodontal destruction. The role of these hormonal changes in pregnant women with different disease patterns remains uncertain. Periodontal health can be reestablished irrespective of the hormonal challenge that takes place during pregnancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226084
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate, 97%
C12H22F6N6OP2