EMAIL THIS PAGE TO A FRIEND
Parasitology Research
The hamster (Mesocricetus auratus) as an experimental model of toxocariasis: histopathological, immunohistochemical, and immunoelectron microscopic findings.
PMID 25518818