EMAIL THIS PAGE TO A FRIEND
Nature
Astrobiology: Prescient words on comets and life.
PMID 25519119
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
31616
Activated charcoal, puriss. p.a., powder
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
675326
Activated charcoal, DARCO® KB-G
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
05112
Activated charcoal, for GC
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
C7606
Activated Charcoal, meets USP testing specifications
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
97876
Activated Charcoal Norit®, Norit CA1, wood, chemically activated, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF41284953
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
C
GF24640950
Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%
C
GF77888121
Carbon, rod, 100mm, diameter 5.0mm, graphite, 100%
C
GF73005825
Carbon, rod, 300mm, diameter 6.35mm, graphite, 100%
C
GF82433538
Carbon, foil, 100x100mm, thickness 0.2mm, flexible graphite, 99.8%
C