EMAIL THIS PAGE TO A FRIEND
Science translational medicine
A rapid antimicrobial susceptibility test based on single-cell morphological analysis.
PMID 25520395
Abstract
A rapid antibiotic susceptibility test (AST) is desperately needed in clinical settings for fast and appropriate antibiotic administration. Traditional ASTs, which rely on cell culture, are not suitable for urgent cases of bacterial infection and antibiotic resistance owing to their relatively long test times. We describe a novel AST called single-cell morphological analysis (SCMA) that can determine antimicrobial susceptibility by automatically analyzing and categorizing morphological changes in single bacterial cells under various antimicrobial conditions. The SCMA was tested with four Clinical and Laboratory Standards Institute standard bacterial strains and 189 clinical samples, including extended-spectrum β-lactamase-positive Escherichia coli and Klebsiella pneumoniae, imipenem-resistant Pseudomonas aeruginosa, methicillin-resistant Staphylococcus aureus, and vancomycin-resistant Enterococci from hospitals. The results were compared with the gold standard broth microdilution test. The SCMA results were obtained in less than 4 hours, with 91.5% categorical agreement and 6.51% minor, 2.56% major, and 1.49% very major discrepancies. Thus, SCMA provides rapid and accurate antimicrobial susceptibility data that satisfy the recommended performance of the U.S. Food and Drug Administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
778362
5-Methyl-5-allyloxycarbonyl-1,3-dioxan-2-one, 97%
C9H12O5