EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Role of Cdk1 in the p53-independent abrogation of the postmitotic checkpoint by human papillomavirus E6.
PMID 25520504
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
81845
Propidium iodide, ≥94% (HPLC)
C27H34I2N4