EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health
Ultrastructural changes in rat Clara cells induced by bis(4-amino-3-methylcyclohexyl)methane.
PMID 2552133
Abstract
Repeated oral administration of bis(4-amino-3-methylcyclohexyl)methane to rats induced unusual ultrastructural changes in Clara cells of the bronchiolar epithelium, involving marked accumulation of electron-dense inclusion bodies with a lamellar structure in the cytoplasm. The appearance of the inclusion bodies was speculated to result from accumulation of complexes formed between the agent and phospholipids within lysosomes, a condition known as drug-induced lipidosis. This finding appeared noteworthy, since it might reflect high potential for metabolism of xenobiotic compounds in Clara cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
369500
4,4′-Methylenebis(2-methylcyclohexylamine), mixture of isomers, 99%
C15H30N2