EMAIL THIS PAGE TO A FRIEND
American journal of reproductive immunology (New York, N.Y. : 1989)
Urinary excretion of C5b-9 is associated with the anti-angiogenic state in severe preeclampsia.
PMID 25521546
Abstract
Severe preeclampsia has been independently linked to complement dysregulation and angiogenic imbalance; however, the relationship between complement and angiogenic factors in human pregnancy is unclear. Utilizing existing biomarkers, our study sought to better understand this relationship in active disease. We performed a case-control study, enrolling 25 cases with severe preeclampsia, 25 controls with chronic hypertension, and 25 healthy controls without hypertension. Levels of complement components (C3a, C5a, and C5b-9) and angiogenic markers [basic fibroblast growth factor (bFGF), placental growth factor (PlGF), vascular endothelial growth factor (VEGF), and soluble fms-like tyrosine kinase-1 (sFlt-1)] were measured simultaneously. Compared to both hypertensive and non-hypertensive controls, severe preeclampsia was associated with increased plasma sFlt-1, decreased plasma VEGF and PlGF, decreased urinary PlGF, and increased urinary C5b-9. Urinary marker C5b-9 correlated strongly with the anti-angiogenic condition. In subjects with detectable urinary excretion of C5b-9, median plasma levels of sFlt-1 were significantly greater (32,029 versus 4556 pg/mL, P < 0.0001) and levels of PlGF (15.6 versus 226 pg/mL, P < 0.0001) and VEGF (119 versus 153 pg/mL, P = 0.001) were significantly lower. More so than plasma complement markers, urinary C5b-9 may a useful measure to link complement dysregulation with angiogenic imbalance in severe preeclampsia.