EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
Field evaluation of picaridin repellents reveals differences in repellent sensitivity between Southeast Asian vectors of malaria and arboviruses.
PMID 25522134