EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Controlled and sustained release of pharmaceuticals via single step solvent-free encapsulation.
PMID 25522209
Abstract
Herein, we report a novel and solvent-free technique for the encapsulation of pharmaceuticals that allows achieving controllable release rates. This method utilizes the deposition of a plasma polymer coating of controlled chemistry and thickness on the outer surface of drug particles placed under continuous agitation.