EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Impairment of social behavior and communication in mice lacking the Uba6-dependent ubiquitin activation system.
PMID 25523030
Abstract
The Uba6-Use1 ubiquitin enzyme cascade is a poorly understood arm of the ubiquitin-proteasome system required for mouse development. Recently, we reported that Uba6 brain-specific knockout (termed NKO) mice display abnormal social behavior and neuronal development due to a decreased spine density and accumulation of Ube3a and Shank3. To better characterize a potential role for NKO mice in autism spectrum disorders (ASDs), we performed a comprehensive behavioral characterization of the social behavior and communication of NKO mice. Our behavioral results confirmed that NKO mice display social impairments, as indicated by fewer vocalizations and decreased social interaction. We conclude that UBA6 NKO mice represent a novel ASD mouse model of anti-social and less verbal behavioral symptoms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30056
Betaine, certified reference material, TraceCERT®
C5H11NO2