EMAIL THIS PAGE TO A FRIEND
Epilepsy Research
Spatiotemporal differences in the c-fos pathway between C57BL/6J and DBA/2J mice following flurothyl-induced seizures: A dissociation of hippocampal Fos from seizure activity.
PMID 25524858
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287571
Bis(2,2,2-trifluoroethyl) ether, 98%
C4H4F6O