EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
De novo design of a transmembrane Zn²⁺-transporting four-helix bundle.
PMID 25525248
Abstract
The design of functional membrane proteins from first principles represents a grand challenge in chemistry and structural biology. Here, we report the design of a membrane-spanning, four-helical bundle that transports first-row transition metal ions Zn(2+) and Co(2+), but not Ca(2+), across membranes. The conduction path was designed to contain two di-metal binding sites that bind with negative cooperativity. X-ray crystallography and solid-state and solution nuclear magnetic resonance indicate that the overall helical bundle is formed from two tightly interacting pairs of helices, which form individual domains that interact weakly along a more dynamic interface. Vesicle flux experiments show that as Zn(2+) ions diffuse down their concentration gradients, protons are antiported. These experiments illustrate the feasibility of designing membrane proteins with predefined structural and dynamic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209988
Zinc, dust, <10 μm, ≥98%
Zn
324930
Zinc, powder, <150 μm, 99.995% trace metals basis
Zn
243469
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zn
31653
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zn
96454
Zinc, purum, powder
Zn
267619
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Zn
578002
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zn
356018
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zn
565148
Zinc, granular, 30-100 mesh, 99%
Zn
267635
Zinc, shot, 5 mm, 99.999% trace metals basis
Zn
267929
Zinc, wire, diam. 1.0 mm, 99.999% trace metals basis
Zn
266361
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Zn
402583
Zinc, shot, <12 mm, 99.99% trace metals basis
Zn
215481
Zinc, mossy, ≥99%
Zn
266353
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zn
GF86667812
Zinc, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95+%
Zn
716723
Zinc, shot, 99.9999%
Zn
349410
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Zn
215503
Zinc, mossy, 99.99% trace metals basis
Zn
GF60963453
Zinc, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.99+%
Zn
GF63340758
Zinc, foil, 100x100mm, thickness 0.20mm, as rolled, 99.95+%
Zn
749060
Zinc, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Zn
GF59993061
Zinc, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95+%
Zn
GF37287704
Zinc, wire reel, 100m, diameter 0.25mm, as drawn, 99.99+%
Zn
GF32078853
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zn
GF72714102
Zinc, wire reel, 0.1m, diameter 1.0mm, 99.999%
Zn
GF47920305
Zinc, wire reel, 10m, diameter 0.125mm, as drawn, 99%
Zn
GF58911385
Zinc, wire reel, 0.5m, diameter 1.0mm, 99.99+%
Zn
GF99245728
Zinc, foil, 300x300mm, thickness 0.20mm, as rolled, 99.95+%
Zn
GF01301612
Zinc, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.9%
Zn